(S)-2-Diphenylphosphino-1-phenylethylamine ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(Diphenylphosphino)-1-phenylethanamine

A721492-1G A721492-250MG
76992-854EA 445.58 USD
76992-854 76992-856
(S)-2-Diphenylphosphino-1-phenylethylamine ≥98%, ee 99%
(S)-2-Diphenylphosphino-1-phenylethylamine

Precaución: for Research Use Only

Formula: C₂₀H₂₀NP
MW: 305.361 g/mol
MDL Number: MFCD17013989
CAS Number: 1103533-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR