5,6-Dimethylpyridin-3-ol 95%

Supplier: Ambeed
Warning

A721235-100MG A721235-1G A721235-250MG
77018-860EA 26.53 USD
77018-860 77018-862 77018-864
5,6-Dimethylpyridin-3-ol 95%
5,6-Dimethylpyridin-3-ol

Precaución: for Research Use Only

Formula: C₇H₉NO
MW: 123.15 g/mol
MDL Number: MFCD12405445
CAS Number: 61893-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR