1-(3-Amino-2,6-dichloropyridin-4-yl)ethanone 97%
Supplier: Ambeed
|
Warning
|
A720822-100MG
A720822-1G
76992-850EA
39
USD
76992-850
76992-852
1-(3-Amino-2,6-dichloropyridin-4-yl)ethanone 97%
1-(3-Amino-2,6-dichloropyridin-4-yl)ethanone
Precaución: for Research Use Only
Formula:
C₇H₆Cl₂N₂O |
MDL Number:
MFCD08235183 CAS Number: 912772-91-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...