4-(4-Fluorophenoxy)aniline 96%

Supplier: Ambeed
Danger

A720734-1G A720734-250MG A720734-5G
77006-124EA 23 USD
77006-124 77006-126 77006-128
4-(4-Fluorophenoxy)aniline 96%
4-(4-Fluorophenoxy)aniline

Precaución: for Research Use Only

Formula: C₁₂H₁₀FNO
MDL Number: MFCD01312350
CAS Number: 36160-82-4
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR