4,5-Difluorophthalonitrile 95%

Supplier: Ambeed
Danger

A719698-100MG A719698-1G A719698-250MG A719698-5G
76992-834EA 13 USD
76992-834 76992-836 76992-838 76992-840
4,5-Difluorophthalonitrile 95%
4,5-Difluorophthalonitrile

Precaución: for Research Use Only

Formula: C₈H₂F₂N₂
MDL Number: MFCD00061281
CAS Number: 134450-56-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR