(R)-Tetrahydrofuran-3-yl 4-methylbenzenesulfonate 97%
Supplier: Ambeed
|
Warning
|
A719470-100MG
A719470-1G
A719470-250MG
A719470-5G
77006-116EA
13.8
USD
77006-116
77006-118
77006-120
77006-122
(R)-Tetrahydrofuran-3-yl 4-methylbenzenesulfonate 97%
(R)-Tetrahydrofuran-3-yl 4-methylbenzenesulfonate
Precaución: for Research Use Only
Formula:
C₁₁H₁₄O₄S MW: 242.29 g/mol |
MDL Number:
MFCD27947543 CAS Number: 219823-47-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...