5-((3,5-Dimethylphenoxy)methyl)oxazolidin-2-one 98%

Supplier: Ambeed
Warning

Synonyms: 5-[(3,5-Xylyloxy)methyl]-2-oxazolidinone, Metaxalone, 5-(3,5-Dimethylphenoxymethyl)-2-oxazolidinone

A718995-100G A718995-100MG A718995-1G A718995-5G
77006-108EA 258.86 USD
77006-108 77006-110 77006-112 77602-840
5-((3,5-Dimethylphenoxy)methyl)oxazolidin-2-one 98%
5-((3,5-Dimethylphenoxy)methyl)oxazolidin-2-one

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₃
MW: 221.25 g/mol
MDL Number: MFCD00867700
CAS Number: 1665-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR