2-(2-Phenyl-2H-tetrazol-5-yl)pyridine 95%

Supplier: Ambeed
Danger

A718466-100MG A718466-1G A718466-250MG
76992-826EA 137.92 USD
76992-826 76992-828 76992-830
2-(2-Phenyl-2H-tetrazol-5-yl)pyridine 95%
2-(2-Phenyl-2H-tetrazol-5-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₉N₅
MW: 223.23 g/mol
CAS Number: 507270-06-6
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR