1-Thiazol-2-yl-ethanol 95%

Supplier: Ambeed
Warning

A718253-1G A718253-250MG
77035-886EA 34 USD
77035-886 77602-838
1-Thiazol-2-yl-ethanol 95%
1-Thiazol-2-yl-ethanol

Precaución: for Research Use Only

Formula: C₅H₇NOS
MDL Number: MFCD13184171
CAS Number: 40982-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR