3-(1H-1,2,4-Triazol-1-yl)benzoic acid 97%

Supplier: Ambeed
Warning

A718155-1G A718155-250MG A718155-5G
77006-102EA 43 USD
77006-102 77006-104 77006-106
3-(1H-1,2,4-Triazol-1-yl)benzoic acid 97%
3-(1H-1,2,4-Triazol-1-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇N₃O₂
MDL Number: MFCD08271895
CAS Number: 167626-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR