3-Bromo-4-(trifluoromethyl)phenol 97%

Supplier: Ambeed
Warning

A718150-10G A718150-5G
76992-824EA 484.84 USD
76992-824 77614-208
3-Bromo-4-(trifluoromethyl)phenol 97%
3-Bromo-4-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₄BrF₃O
MW: 241.01 g/mol
MDL Number: MFCD13194488
CAS Number: 1214385-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR