Methyl-2-benzyloxycarbonylamino-3-hydroxypropionate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(((benzyloxy)carbonyl)amino)-3-hydroxypropanoate

A718037-100G A718037-10G A718037-1G A718037-25G A718037-5G
77006-092EA 727.78 USD
77006-092 77006-094 77006-096 77006-098 77006-100
Methyl-2-benzyloxycarbonylamino-3-hydroxypropionate 97%
Methyl-2-benzyloxycarbonylamino-3-hydroxypropionate

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₅
MW: 253.26 g/mol
MDL Number: MFCD00136653
CAS Number: 14464-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR