6-Methylthiazolo[4,5-b]pyridine-2-thiol 98%

Supplier: Ambeed
Warning

A717924-1G A717924-5G
76992-820EA 42.44 USD
76992-820 76992-822
6-Methylthiazolo[4,5-b]pyridine-2-thiol 98%
6-Methylthiazolo[4,5-b]pyridine-2-thiol

Precaución: for Research Use Only

Formula: C₇H₆N₂S₂
MW: 182.27 g/mol
MDL Number: MFCD13195716
CAS Number: 586409-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR