2-(4-(2,5-Dimethyl-1H-pyrrol-1-yl)phenyl)acetic acid 97%

Supplier: Ambeed
Warning

A717848-1G A717848-250MG
77006-088EA 63 USD
77006-088 77006-090
2-(4-(2,5-Dimethyl-1H-pyrrol-1-yl)phenyl)acetic acid 97%
2-(4-(2,5-Dimethyl-1H-pyrrol-1-yl)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO₂
MDL Number: MFCD01874280
CAS Number: 26165-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR