(3-Chloro-5-(trifluoromethyl)phenyl)acetic acid 98%

Supplier: Ambeed
Warning

A717809-1G A717809-250MG A717809-5G A717809-10G
77035-880EA 44 USD
77035-880 77035-882 77035-884 77873-130
(3-Chloro-5-(trifluoromethyl)phenyl)acetic acid 98%
(3-Chloro-5-(trifluoromethyl)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O₂
MDL Number: MFCD04115851
CAS Number: 886496-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR