2,6-Di(1H-imidazol-1-yl)pyridine 98%
Supplier: Ambeed
|
Warning
|
A717628-1G
A717628-250MG
A717628-5G
77018-830EA
72
USD
77018-830
77018-832
77018-834
2,6-Di(1H-imidazol-1-yl)pyridine 98%
2,6-Di(1H-imidazol-1-yl)pyridine
Precaución: for Research Use Only
Formula:
C₁₁H₉N₅ MW: 211.22 g/mol |
MDL Number:
MFCD23103190 CAS Number: 39242-17-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...