N-[(1,3-Dimethyl-1H-pyrazol-5-yl)methyl]-N-methylamine 95%

Supplier: Ambeed
Warning

Synonyms: 1-(1,3-Dimethyl-1H-pyrazol-5-yl)-N-methylmethanamine

A717465-1G A717465-250MG
77006-084EA 78.51 USD
77006-084 77006-086
N-[(1,3-Dimethyl-1H-pyrazol-5-yl)methyl]-N-methylamine 95%
N-[(1,3-Dimethyl-1H-pyrazol-5-yl)methyl]-N-methylamine

Precaución: for Research Use Only

Formula: C₇H₁₃N₃
MW: 139.2 g/mol
MDL Number: MFCD08700823
CAS Number: 852227-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR