Methyl 3-chloro-5-formylbenzoate 98%

Supplier: Ambeed
Warning

A717362-250MG A717362-100MG
77018-828EA 123.07 USD
77018-828 77614-206
Methyl 3-chloro-5-formylbenzoate 98%
Methyl 3-chloro-5-formylbenzoate

Precaución: for Research Use Only

Formula: C₉H₇ClO₃
MW: 198.60 g/mol
MDL Number: MFCD22544335
CAS Number: 879542-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR