Methyl-3-bromo-4-methoxybenzoate 98%

Supplier: Ambeed
Danger

A717027-1G A717027-25G A717027-5G
77035-874EA 13 USD
77035-874 77035-876 77035-878
Methyl-3-bromo-4-methoxybenzoate 98%
Methyl-3-bromo-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MDL Number: MFCD00210463
CAS Number: 35450-37-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR