2-(1-Hydroxyethyl)benzimidazole 95%

Supplier: Ambeed
Warning

A716615-100G A716615-25G A716615-5G A716615-10G
77006-070EA 381 USD
77006-070 77006-072 77006-074 77673-968
2-(1-Hydroxyethyl)benzimidazole 95%
2-(1-Hydroxyethyl)benzimidazole

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O
MDL Number: MFCD00016718
CAS Number: 19018-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR