L-Methionine sulfone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-2-Amino-4-(methylsulphonyl)butanoic acid
A715977-1G
A715977-250MG
A715977-25G
A715977-5G
76992-804EA
26
USD
76992-804
76992-806
76992-808
76992-810
L-Methionine sulfone 98%
L-Methionine sulphone
Precaución: for Research Use Only
Formula:
C₅H₁₁NO₄S MW: 181.21 g/mol |
MDL Number:
MFCD00066020 CAS Number: 7314-32-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...