1-Chloro-3-methylbutan-2-one 95%

Supplier: Ambeed
Danger

A715864-100MG A715864-1G A715864-250MG
76992-798EA 127.31 USD
76992-798 76992-800 76992-802
1-Chloro-3-methylbutan-2-one 95%
1-Chloro-3-methylbutan-2-one

Precaución: for Research Use Only

Formula: C₅H₉ClO
MW: 120.57 g/mol
MDL Number: MFCD19160714
CAS Number: 17687-63-7
UN: 1992
ADR: 3,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR