2-Bromo-4,6-diphenylpyrimidine 98%

Supplier: Ambeed
Warning

A715787-100MG A715787-1G
77018-820EA 7 USD
77018-820 77018-822
2-Bromo-4,6-diphenylpyrimidine 98%
2-Bromo-4,6-diphenylpyrimidine

Precaución: for Research Use Only

Formula: C₁₆H₁₁BrN₂
MDL Number: MFCD00234884
CAS Number: 56181-49-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR