1-Ethyl-1H-indole-5-carbaldehyde 95%

Supplier: Ambeed
Warning

A714518-1G A714518-250MG A714518-5G
77035-858EA 89.12 USD
77035-858 77035-860 77035-862
1-Ethyl-1H-indole-5-carbaldehyde 95%
1-Ethyl-1H-indole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO
MDL Number: MFCD09455244
CAS Number: 944893-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR