6-fluoro-4-methylpyridin-3-amine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoro-4-methylpyridin-3-amine, 5-Amino-2-fluoro-4-methylpyridine

A714201-1G A714201-250MG A714201-25G A714201-5G
77006-060EA 30.77 USD
77006-060 77006-062 77006-064 77006-066
6-fluoro-4-methylpyridin-3-amine 97%
6-fluoro-4-methylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₇FN₂
MW: 126.134 g/mol
MDL Number: MFCD09864696
CAS Number: 954236-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR