3-Cyano-5-methoxyphenylboronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: 3-Cyano-5-methoxyphenylboronic acid, pinacol ester, 3-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

A713262-100MG A713262-1G A713262-250MG
77035-840EA 37.14 USD
77035-840 77035-842 77035-844
3-Cyano-5-methoxyphenylboronic acid pinacol ester 96%
3-Cyano-5-methoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BNO₃
MW: 259.11 g/mol
MDL Number: MFCD11855976
CAS Number: 1035266-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR