2-Amino-3-methylpyridine-5-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine

A713214-100MG A713214-1G A713214-250MG A713214-5G
77035-834EA 39 USD
77035-834 77035-836 77035-838 77873-120
2-Amino-3-methylpyridine-5-boronic acid pinacol ester 98%
2-Amino-3-methylpyridine-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₉BN₂O₂
MDL Number: MFCD12923388
CAS Number: 1111637-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR