Imidazo[1,2-a]pyridine-7-methanol 96%

Supplier: Ambeed
Warning

A712335-100MG A712335-1G A712335-250MG A712335-5G
77018-810EA 19.1 USD
77018-810 77018-812 77614-198 77684-632
Imidazo[1,2-a]pyridine-7-methanol 96%
Imidazo[1,2-a]pyridine-7-methanol

Precaución: For research use only.

Formula: C₈H₈N₂O
MW: 148.16 g/mol
MDL Number: MFCD12400828
CAS Number: 342613-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR