Imidazo[1,2-a]pyridine-2-carbaldehyde 96%

Supplier: Ambeed
Warning

Synonyms: Imidazo[1,2-a]pyridine-2-carboxaldehyde

A712105-1G A712105-250MG A712105-5G A712105-100MG
77018-804EA 87 USD
77018-804 77018-806 77018-808 77684-631
Imidazo[1,2-a]pyridine-2-carbaldehyde 96%
Imidazo[1,2-a]pyridine-2-carbaldehyde

Precaución: For research use only.

Formula: C₈H₆N₂O
MW: 146.1460 g/mol
MDL Number: MFCD06739247
CAS Number: 118000-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR