5,6-Dichloro[1,2,5]oxadiazolo[3,4-b]pyrazine 97%

Supplier: Ambeed
Warning

Synonyms: 5,6-Dichloro-[1,2,5]oxadiazolo[3,4-b]pyrazine

A712083-250MG A712083-5G A712083-1G
77006-056EA 112.46 USD
77006-056 77006-058 77673-952
5,6-Dichloro[1,2,5]oxadiazolo[3,4-b]pyrazine 97%
5,6-Dichloro[1,2,5]oxadiazolo[3,4-b]pyrazine

Precaución: for Research Use Only

Formula: C₄Cl₂N₄O
MW: 190.98 g/mol
MDL Number: MFCD00187160
CAS Number: 153493-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR