3-(4-Cyanophenylcarbamoyl)phenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (3-((4-Cyanophenyl)carbamoyl)phenyl)boronic acid

A711970-1G A711970-250MG A711970-5G
77018-798EA 62.6 USD
77018-798 77018-800 77018-802
3-(4-Cyanophenylcarbamoyl)phenylboronic acid 95%
3-(4-Cyanophenylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₁BN₂O₃
MW: 266.0600 g/mol
MDL Number: MFCD06659886
CAS Number: 850567-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR