2-Amino-3,3,3-trifluoropropionic acid ethylester hydrochloride 98%

Supplier: Ambeed
Warning

A711650-1G A711650-250MG A711650-5G
77035-826EA 105 USD
77035-826 77035-828 77035-830
2-Amino-3,3,3-trifluoropropionic acid ethylester hydrochloride 98%
2-Amino-3,3,3-trifluoropropionic acid ethylester hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉ClF₃NO₂
MDL Number: MFCD14525479
CAS Number: 193140-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR