2-Hydroxy-5-(trifluoromethyl)benzaldehyde 95%

Supplier: Ambeed
Warning

A711117-100MG A711117-1G A711117-250MG A711117-5G
76992-762EA 13 USD
76992-762 76992-764 76992-766 76992-768
2-Hydroxy-5-(trifluoromethyl)benzaldehyde 95%
2-Hydroxy-5-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₅F₃O₂
MDL Number: MFCD16999709
CAS Number: 210039-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR