Ethyl 4-Bromo-3-methylbenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-bromo-3-methylbenzoate, 4-Bromo-m-toluic acid Ethyl ester, 4-Bromo-3-methylbenzoic acid Ethyl ester, Ethyl 4-Bromo-m-toluate

A711054-100G A711054-1G A711054-25G A711054-5G
77035-818EA 172.93 USD
77035-818 77035-820 77035-822 77035-824
Ethyl 4-Bromo-3-methylbenzoate 98%
Ethyl 4-Bromo-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MW: 243.10 g/mol
MDL Number: MFCD00673007
CAS Number: 160313-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR