1-Decanol 98%

Supplier: Ambeed
Warning

Synonyms: 1-Hydroxy decane, 1-Decyl alcohol, Capric alcohol, Decan-1-ol, Nonyl carbinol, Decyl alcohol

A710792-1000G A710792-100G A710792-25G A710792-500G
77006-048EA 29 USD
77006-048 77006-050 77006-052 77006-054
1-Decanol 98%
1-Decanol

Precaución: for Research Use Only

Formula: H₃C(CH₂)₈CH₂OH
MW: 158.28 g/mol
Boiling Pt: 230
Melting Pt: 7
Flash Pt: 82
MDL Number: MFCD00004747
CAS Number: 112-30-1
Merck Index: 13,02875

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR