2-(Iodomethyl)Oxetane 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A710602-1G A710602-250MG
76992-756EA 160 USD
76992-756 77684-626
2-(Iodomethyl)Oxetane 98%, Ambeed.Inc
2-(Iodomethyl)Oxetane

Precaución: For research use only.

Formula: C₄H₇IO
MDL Number: MFCD11226849
CAS Number: 121138-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR