Methyl 2-bromo-2-(4-methoxyphenyl)acetate 95%

Supplier: Ambeed
Danger

A709832-1G A709832-250MG A709832-5G
77018-796EA 105 USD
77018-796 77891-395 77891-396
Methyl 2-bromo-2-(4-methoxyphenyl)acetate 95%
Methyl 2-bromo-2-(4-methoxyphenyl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₃
MDL Number: MFCD11847980
CAS Number: 50612-99-2
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR