4-Chloro-6-isopropylpyrimidine 95%

Supplier: Ambeed
Warning

A708481-100MG A708481-1G A708481-250MG
76992-748EA 113 USD
76992-748 76992-750 76992-752
4-Chloro-6-isopropylpyrimidine 95%
4-Chloro-6-isopropylpyrimidine

Precaución: for Research Use Only

Formula: C₇H₉ClN₂
MDL Number: MFCD09750199
CAS Number: 954222-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR