4-Bromo-3-cyanobenzene-1-sulfonyl chloride 95%

Supplier: Ambeed
Danger

A708343-100MG A708343-250MG
76992-744EA 92.3 USD
76992-744 76992-746
4-Bromo-3-cyanobenzene-1-sulfonyl chloride 95%
4-Bromo-3-cyanobenzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₇H₃BrClNO₂S
MW: 280.53 g/mol
MDL Number: MFCD18390733
CAS Number: 1261583-46-3
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR