2-Fluoro-4-iodo-3,5-dimethylpyridine 95%

Supplier: Ambeed
Warning

A707856-100MG A707856-250MG
77018-790EA 83.82 USD
77018-790 77614-180
2-Fluoro-4-iodo-3,5-dimethylpyridine 95%
2-Fluoro-4-iodo-3,5-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₇FIN
MW: 251.04 g/mol
CAS Number: 1429510-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR