6-Chloro-5-methylpyrimidin-4-ol 95%

Supplier: Ambeed
Danger

A707717-100MG A707717-250MG A707717-5G A707717-1G
77006-032EA 22.28 USD
77006-032 77006-034 77602-808 77673-940
6-Chloro-5-methylpyrimidin-4-ol 95%
6-Chloro-5-methylpyrimidin-4-ol

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O
MDL Number: MFCD09879261
CAS Number: 18261-42-2
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR