4-(1H-Pyrazol-4-yl)-7-((2-(trimethylsilyl)ethoxy)methyl)-7H-pyrrolo[2,3-d]pyrimidine 99+%

Supplier: Ambeed
Warning

A707673-10MG A707673-1G A707673-1MG A707673-250MG A707673-25MG A707673-50MG A707673-5G
77006-018EA 7 USD
77006-018 77006-020 77006-022 77006-024 77006-026 77006-028 77006-030
4-(1H-Pyrazol-4-yl)-7-((2-(trimethylsilyl)ethoxy)methyl)-7H-pyrrolo[2,3-d]pyrimidine 99+%
4-(1H-Pyrazol-4-yl)-7-((2-(trimethylsilyl)ethoxy)methyl)-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₁₅H₂₁N₅OSi
MDL Number: MFCD11857754
CAS Number: 941685-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR