(5-Methyl-1,3,4-oxadiazol-2-yl)methanol 96%

Supplier: Ambeed
Warning

A707396-100MG A707396-250MG A707396-5G A707396-1G
77018-784EA 70.02 USD
77018-784 77018-786 77018-788 77573-296
(5-Methyl-1,3,4-oxadiazol-2-yl)methanol 96%
(5-Methyl-1,3,4-oxadiazol-2-yl)methanol

Precaución: for Research Use Only

Formula: C₄H₆N₂O₂
MW: 114.1026 g/mol
MDL Number: MFCD08059900
CAS Number: 915924-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR