Methyl 2-bromo-6-methoxynicotinate 96%

Supplier: Ambeed
Warning

A707143-100MG A707143-1G A707143-250MG
77018-778EA 32 USD
77018-778 77018-780 77018-782
Methyl 2-bromo-6-methoxynicotinate 96%
Methyl 2-bromo-6-methoxynicotinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
MDL Number: MFCD18257522
CAS Number: 1009735-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR