3,6-Dibromo-9-(2-ethylhexyl)carbazole 98%

Supplier: Ambeed
Warning

A706805-250MG A706805-5G
76992-728EA 33 USD
76992-728 76992-730
3,6-Dibromo-9-(2-ethylhexyl)carbazole 98%
3,6-Dibromo-9-(2-ethylhexyl)carbazole

Precaución: for Research Use Only

Formula: C₂₀H₂₃Br₂N
MDL Number: MFCD18252907
CAS Number: 173063-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR