1H-Pyrrolo[2,3-b]pyridine-2-carboxaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 1H-Pyrrolo[2,3-b]pyridine-2-carbaldehyde

A705291-1G A705291-250MG A705291-5G
76992-712EA 151.71 USD
76992-712 76992-714 76992-716
1H-Pyrrolo[2,3-b]pyridine-2-carboxaldehyde 97%
1H-Pyrrolo[2,3-b]pyridine-2-carboxaldehyde

Precaución: for Research Use Only

Formula: C₈H₆N₂O
MW: 146.15 g/mol
MDL Number: MFCD08437634
CAS Number: 394223-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR