4,4'-Stilbenedicarboxylic acid 95%

Supplier: Ambeed
Warning

A705140-100G A705140-1G A705140-5G
77035-802EA 99 USD
77035-802 77035-804 77035-806
4,4'-Stilbenedicarboxylic acid 95%
4,4'-Stilbenedicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₂O₄
MDL Number: MFCD00013994
CAS Number: 100-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR