2-Methyl-[1,1'-biphenyl]-4,4'-dicarboxylic acid 96%

Supplier: Ambeed
Warning

A704541-100MG A704541-1G A704541-250MG
76992-704EA 33.95 USD
76992-704 76992-706 76992-708
2-Methyl-[1,1'-biphenyl]-4,4'-dicarboxylic acid 96%
2-Methyl-[1,1'-biphenyl]-4,4'-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₂O₄
MW: 256.25 g/mol
MDL Number: MFCD18320269
CAS Number: 1186048-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR