2-Oxo-2-phenylacetaldehyde hydrate 98%

Supplier: Ambeed
Warning

A704490-1G A704490-5G
77018-770EA 13 USD
77018-770 77893-416
2-Oxo-2-phenylacetaldehyde hydrate 98%
2-Oxo-2-phenylacetaldehyde hydrate

Precaución: for Research Use Only

Formula: C₈H₆O₂·H₂O
MDL Number: MFCD00149499
CAS Number: 78146-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR