2-(3,5-Dimethylphenyl)quinoline 97%

Supplier: Ambeed
Warning

A703957-1G A703957-5G
77018-758EA 30 USD
77018-758 77018-760
2-(3,5-Dimethylphenyl)quinoline 97%
2-(3,5-Dimethylphenyl)quinoline

Precaución: for Research Use Only

Formula: C₁₇H₁₅N
MW: 233.30 g/mol
MDL Number: MFCD22571707
CAS Number: 1056451-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR